missing translation for 'onlineSavingsMsg'
Learn More

NUMBL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-88136

Product Code. 18109009

  • € 570.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NUMBL Polyclonal specifically detects NUMBL in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUMBL
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
CAG3A, CTG3a, numb (Drosophila) homolog-like, numb homolog (Drosophila)-like, numb homolog-like, NUMBLIKE, numb-like protein, NUMB-R, Numb-related protein, NUMBRNBL, TNRC23
Rabbit
Affinity Purified
RUO
9253
Human
IgG
Immunoprecipitation
1.0 mg/mL
Immunoprecipitation 2 - 5 ug/mg lysate
NP_004747.1
NUMBL
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human numb (Drosophila) homolog-like using the numbering given in entry NP_004747.1 (GeneID 9253).
100 μL
Primary
Based on 100% sequence identity, this antibody is predicted to react with Pig, Orangutan, Rhesus Monkey, Chimpanzee and Crab-eating macaque.
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only