missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMBL Monoclonal Antibody (OTI8D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812202S
This item is not returnable.
View return policy
Description
NUMBL Monoclonal antibody specifically detects NUMBL in Human samples. It is validated for Western BlotSpecifications
| NUMBL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14061 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10063 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8D2 | |
| Unconjugated | |
| COX17 | |
| CAG3A; CTG3a; NBL; numb homolog (Drosophila)-like; numb homolog-like; NUMB like, endocytic adaptor protein; Numbl; NUMBLIKE; numb-like; numb-like protein; NUMBR; Numb-R; numb-related protein; TNRC23 | |
| Human recombinant protein fragment corresponding to amino acids 2-63 of human COX17 (NP_005685) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction