missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMBL Monoclonal Antibody (OTI8D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812202
This item is not returnable.
View return policy
Description
The mammalian NUMB homolog plays a role in the determination of cell fate during development and binds with a variety of proteins, including Eps15, LNX1 and Notch 1. NumbL (NUMB-like protein), also known as Numb-R, NBL, CAG3A, CTG3a, NUMBLIKE or TNRC23, is a 609 amino acid cytoplasmic protein that, like NUMB, is thought to play a role in cell fate. Expressed at high levels in developing brain tissue, NumbL contains one PID (phosphotyrosine interaction domain) and plays an important role in neuronal differentiation, possibly associating with Eps15 and Notch 1. In mice, deletion of the NumbL gene is associated with early embryonic death, suggesting an essential role for NumbL in early development.Specifications
| NUMBL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6R0 | |
| NUMBL | |
| Human recombit protein fragment corresponding to amino acids 1-225 of human NUMBL produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8D2 | |
| Unconjugated | |
| NUMBL | |
| CAG3A, CTG3a, NBL, NUMB-R, NUMBLIKE, NUMBR, TNRC23 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9253 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction