missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUP133 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17762614
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17762614 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17762614 Supplier Invitrogen™ Supplier No. PA5121218

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A375, LO2, HeLa, NCI-H460. The target is usually found in the following locations: Chromosome, Nucleus, centromere, kinetochore, nuclear pore complex. Immunogen sequence: NILKDMLQAA SHYRQNRNSL YRREESLEKE PEYVPWTATS GPGGIRTVII RQHEIVLKVA YPQADSNLRN IVTEQLVALI DCFLDGYVSQ LKSVDKSSNR ERYDNLEMEY LQKRSDLLSP LLSLGQYLWA ASLAEKYCDF DILVQMCEQT DNQSRLQRYM TQFADQNFSD FLFRWYLEKG KRGKLLSQPI SQHGQLANFL Q.

Plays a role in T-cell signaling pathway. Isoform 2 may play a role in T-helper 2 (Th2) signaling pathway and seems to represent the first proximal signaling protein that links T-cell receptor-mediated signal to the activation of c-Maf Th2 specific factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP133
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NUP133
Gene Accession No. Q8WUM0
Gene Alias 133 kDa nucleoporin; hNUP133; mermaid; Nuclear pore complex protein Nup133; nucleoporin 133; nucleoporin 133kD; nucleoporin 133kDa; nucleoporin Nup133; NUP133
Gene Symbols NUP133
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 730-930 of human NUP133 (NP_060700.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55746
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.