missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP205 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22287PE
This item is not returnable.
View return policy
Description
NUP205 Polyclonal antibody specifically detects NUP205 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| NUP205 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| nucleoporin 205kDa | |
| The immunogen this antibody was made to, maps to a region between residue 1750 and 1800 of human Nucleoporin 205kDa using the numbering given in entry NP_055950.1 (GeneID 23165). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23165 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction