missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP205 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24439-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a nucleoporin, which is a subunit of the nuclear pore complex that functions in active transport of proteins, RNAs and ribonucleoprotein particles between the nucleus and cytoplasm. Mutations in this gene are associated with steroid-resistant nephrotic syndrome.Specifications
| NUP205 | |
| Polyclonal | |
| Unconjugated | |
| NUP205 | |
| 205 kDa nucleoporin, C7orf14, KIAA0225, nucleoporin 205kDa, Nucleoporin Nup205, NUP205 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23165 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92621 | |
| NUP205 | |
| NUP205 Fusion Protein Ag19832 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction