missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP210 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-93336
This item is not returnable.
View return policy
Description
NUP210 Polyclonal specifically detects NUP210 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| NUP210 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| FLJ22389, GP210Pore membrane protein of 210 kDa, KIAA0906Nuclear pore protein gp210, Nuclear envelope pore membrane protein POM 210, nuclear pore membrane glycoprotein 210, nucleoporin 210kDa, POM210Nucleoporin Nup210 | |
| Rabbit | |
| 210 kDa | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, Small-eared galago and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q8TEM1 | |
| NUP210 | |
| The immunogen recognized by this antibody maps to a region between residue 1837 and 1887 of human nucleoporin 210kDa using the numbering given in entry NP_079199.2 (GeneID 23225). | |
| Affinity Purified | |
| RUO | |
| 23225 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction