missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP210 Mouse anti-Human, Clone: OTI7G1, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812297
This item is not returnable.
View return policy
Description
NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.Specifications
| NUP210 | |
| Monoclonal | |
| Unconjugated | |
| NUP210 | |
| GP210, POM210 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23225 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI7G1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TEM1 | |
| NUP210 | |
| Human recombit protein fragment corresponding to amino acids 27-400 of human NUP210 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction