missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP214 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2244
This item is not returnable.
View return policy
Description
NUP214 Polyclonal specifically detects NUP214 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| NUP214 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| 214 kDa nucleoporin, CAINputative oncogene, D9S46Enuclear pore complex protein Nup214, KIAA0023, MGC104525, N214, nucleoporin 214kD (CAIN), nucleoporin 214kDa, Nucleoporin Nup214, p250, Protein CAN | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8021 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:100- 1:500 | |
| NP_005076.3 | |
| NUP214 | |
| The immunogen recognized by this antibody maps to a region between residue 1250 and 1300 of human Nuceloporin 214 using the numbering given in entry NP_005076.3 (GeneID 8021). | |
| 0.1 mL | |
| Primary | |
| NUP214 - Nuclear Marker. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction