missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP43 Antibody (OTI6F8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73131FR
This item is not returnable.
View return policy
Description
NUP43 Monoclonal specifically detects NUP43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| NUP43 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| NUP43 | |
| Recombinant protein fragment corresponding to amino acids 1-253 of human NUP43 (NP_942590) produced in E.coli. | |
| 0.1 mL | |
| 348995.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6F8 | |
| Western Blot, Immunohistochemistry | |
| bA350J20.1, FLJ13287, nucleoporin 43kDa, nucleoporin Nup43, Nup107-160 subcomplex subunit Nup43, p42 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction