Learn More
Novus Biologicals™ NUP93 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29696
Product Code. 30047789
  • € 571.00 / 100µg
Description
NUP93 Polyclonal antibody specifically detects NUP93 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NUP93
Polyclonal
Tris-citrate/phosphate buffer, pH 7 to 8
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation, Western Blot
Unconjugated
KIAA0095Nucleoporin Nup93,93 kDa nucleoporin, MGC21106, nuclear pore complex protein Nup93, nucleoporin 93kDa
Between 606 and 656
100 μg
Signal Transduction
9688
Store at 4°C. Do not freeze.