Learn More
Novus Biologicals™ NUP93 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29696
This item is not returnable.
View return policy
Description
NUP93 Polyclonal antibody specifically detects NUP93 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NUP93 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| KIAA0095Nucleoporin Nup93,93 kDa nucleoporin, MGC21106, nuclear pore complex protein Nup93, nucleoporin 93kDa | |
| Between 606 and 656 | |
| 100 μg | |
| Signal Transduction | |
| 9688 | |
| Store at 4°C. Do not freeze. | |
| IgG |