missing translation for 'onlineSavingsMsg'
Learn More

Nur77 Mouse anti-Mouse, Human, Alexa Fluor™ 647, Clone: 12.14, BD Pharmingen™

Product Code. 17237564
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17237564 0.1 mg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17237564 Supplier BD Biosciences Supplier No. 566735

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nur77 Monoclonal antibody specifically detects Nur77 in Mouse,Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nur77
Applications Flow Cytometry
Classification Monoclonal
Clone 12.14
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Host Species Mouse
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Nur77
Target Species Mouse, Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.