missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nurim Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA141391

Product Code. 11584400

  • € 530.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Suggested positive control: HeLa, NIH-3T3, PC3. Predicted to react with Chimpanzee and Rhesus Monkey.

Nurim is an inner nuclear membrane (INM) protein that is tightly associated with the inner nuclear membrane. It was first isolated in a visual screen for nuclear envelope-localizing proteins. In subcellular fractionation, nurim remained extremely tightly bound to nuclear fractions. Unlike the known nuclear envelope (NE) membrane proteins, it is neither associated with nuclear pores, nor targeted like lamin-associated membrane proteins suggesting its localization to NE by a distinct mechanism.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nurim
Polyclonal
Unconjugated
NRM
2610307M02Rik; AI429796; DAAP-285E11.8; Nrm; NRM29; Nuclear envelope membrane protein; nuclear rim protein; Nurim; nurim (nuclear envelope membrane protein); UNQ555/PRO1112
Rabbit
Protein G
RUO
106582, 11270
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q8IXM6, Q8VC65
NRM
A portion of amino acids 200-250 of human Nurim.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.