missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUT Antibody (2B6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31147H
This item is not returnable.
View return policy
Description
NUT Monoclonal specifically detects NUT in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NUT | |
| Monoclonal | |
| HRP | |
| C15orf55, chromosome 15 open reading frame 55, DKFZp434O192, FAM22H, MGC138683, MGC138684, Nuclear protein in testis, NUT, protein NUT | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2B6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NUTM1 | |
| Partial recombinant protein made to an internal portion of human NUT (between amino acids 400-700) [UniProt Q86Y26]. | |
| 0.1 mL | |
| Cancer | |
| 256646 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction