missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUT Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32672
This item is not returnable.
View return policy
Description
NUT Monoclonal antibody specifically detects NUT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| NUT | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:800, Immunohistochemistry-Paraffin 1:1000-1:20000 | |
| C15orf55, chromosome 15 open reading frame 55, DKFZp434O192, FAM22H, MGC138683, MGC138684, Nuclear protein in testis, NUT, protein NUT | |
| Recombinant protein within C terminal Mouse NUT. (Uniprot: Q86Y26) | |
| 100 μL | |
| Biologically Active Proteins, Cancer, Immunology, Inflammation | |
| 256646 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction