Learn More
Invitrogen™ NUTF2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101729
Product Code. 16648204
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total NUTF2.
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NUTF2
Polyclonal
Unconjugated
NUTF2
2700067I02Rik; AI115459; AI413596; AW546000; Ntf2; NTF-2; Ntf3; nuclear transport factor 2; NUTF2; Placental protein 15; PP15
Rabbit
Affinity chromatography
RUO
10204, 291981, 68051
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P61970, P61971, P61972
NUTF2
A synthesized peptide derived from human NUTF2(Accession P61970), corresponding to amino acid residues D25-P75.