Learn More
Nutlin-3b, MedChemExpress
Product Code. 16489047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16489047
Brand: Medchem Express HY15335
MedChemExpress Nutlin-3b is a p53/MDM2 inhibitor with an IC50 of 13.6 μM. Nutlin-3b is 150 times less potent in binding to MDM2 than Nutlin-3a.
Nutlin-3b is a p53/MDM2 inhibitor with an IC50 of 13.6 μM. Nutlin-3b is 150 times less potent in binding to MDM2 than Nutlin-3a.
Specifications
Chemical Name or Material Nutlin-3b
CAS 675576-97-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H30Cl2N4O4
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (171.97 mM)
SMILES O=C1NCCN(C(N2[C@@H](C3=CC=C(Cl)C=C3)[C@@H](C4=CC=C(Cl)C=C4)N=C2C5=CC=C(OC)C=C5OC(C)C)=O)C1
Molecular Weight (g/mol) 581.49
Formula Weight 581.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only