missing translation for 'onlineSavingsMsg'
Learn More

Nutlin-3b, MedChemExpress

Product Code. 16482557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16482557

Brand: Medchem Express HY15335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nutlin-3b is a p53/MDM2 inhibitor with an IC50 of 13.6 μM. Nutlin-3b is 150 times less potent in binding to MDM2 than Nutlin-3a.

Nutlin-3b is a p53/MDM2 inhibitor with an IC50 of 13.6 μM. Nutlin-3b is 150 times less potent in binding to MDM2 than Nutlin-3a.

Specifications

Chemical Name or Material Nutlin-3b
CAS 675576-97-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H30Cl2N4O4
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (171.97 mM)
SMILES O=C1NCCN(C(N2[C@@H](C3=CC=C(Cl)C=C3)[C@@H](C4=CC=C(Cl)C=C4)N=C2C5=CC=C(OC)C=C5OC(C)C)=O)C1
Molecular Weight (g/mol) 581.49
Formula Weight 581.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.