Learn More
Thermo Scientific™ Nutrient Broth (Dehydrated)
Description
Cultivate fastidious pathogens and other microorganisms with nutritious Thermo Scientific™ Oxoid™ Nutrient Broth (Dehydrated). This general purpose medium contains beef extracts combined with peptone, glucose, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
A general purpose fluid medium for the cultivation of microorganisms not exacting in their nutritional requirements.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Nutrient Broth |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 38.5L medium |
For Laboratory Use Only