missing translation for 'onlineSavingsMsg'
Learn More

NVP-2, MedChemExpress

Product Code. 16467747
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467747 5 mg 5mg
16491257 50 mg 50mg
16441229 1 mg 1mg
16405028 10 mg 10mg
16425776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16467747 Supplier Medchem Express Supplier No. HY12214A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-2 is a potent and selective ATP-competitive cyclin dependent kinase 9 (CDK9) probe, inhibits CDK9/CycT activity with an IC50 of 0.514 nM. NVP-2 displays inhibitory effcts on CDK1/CycB, CDK2/CycA and CDK16/CycY kinases with IC50 values of 0.584 μM, 0.706 μM, and 0.605 μM, respectively. NVP-2 induces cell apoptosis.

NVP-2 is a potent and selective ATP-competitive cyclin dependent kinase 9 (CDK9) probe, inhibits CDK9/CycT activity with an IC50 of 0.514 nM. NVP-2 displays inhibitory effcts on CDK1/CycB, CDK2/CycA and CDK16/CycY kinases with IC50 values of 0.584 μM, 0.706 μM, and 0.605 μM, respectively. NVP-2 induces cell apoptosis.

Specifications

Chemical Name or Material NVP-2
CAS 1263373-43-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H37ClN6O2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (194.91 mM)
SMILES ClC1=CN=C(N[C@H]2CC[C@H](N[C@H](C)COC)CC2)C=C1C3=CC=CC(NCC4(C#N)CCOCC4)=N3
Molecular Weight (g/mol) 513.07
Formula Weight 513.07
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.