missing translation for 'onlineSavingsMsg'
Learn More

NVP-CGM097 sulfate, MedChemExpress

Product Code. 16452987
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452987

Brand: Medchem Express HY15954B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-CGM097 sulfate is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2.

NVP-CGM097 sulfate is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2.

Specifications

Chemical Name or Material NVP-CGM097 sulfate
CAS 1313367-56-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C38H49ClN4O8S
Quantity 50 mg
Synonym CGM097 sulfate
Solubility Information DMSO : 100 mg/mL (132.04 mM; Need ultrasonic) ∣H2O : 100 mg/mL (132.04 mM; Need ultrasonic)
SMILES ClC(C=C1)=CC=C1[C@@H](C2=CC(OC(C)C)=C(OC)C=C2C3)N(C4=CC=C(N(C[C@@H]5CC[C@@H](N6CC(N(C)CC6)=O)CC5)C)C=C4)C3=O.O=S(O)(O)=O
Molecular Weight (g/mol) 757.34
Formula Weight 757.34
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.