Learn More
NVP-TAE 684, MedChemExpress
Product Code. 16490027
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16490027
Brand: Medchem Express HY10192
MedChemExpress NVP-TAE 684 (TAE 684) is a highly potent and selective ALK inhibitor, which blocks the growth of ALCL-derived and ALK-dependent cell lines with IC50 values between 2 and 10 nM.
NVP-TAE 684 (TAE 684) is a highly potent and selective ALK inhibitor, which blocks the growth of ALCL-derived and ALK-dependent cell lines with IC50 values between 2 and 10 nM.
Specifications
Chemical Name or Material NVP-TAE 684
CAS 761439-42-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H40ClN7O3S
Quantity 50 mg
Synonym TAE 684
Solubility Information DMSO : 7.69 mg/mL (12.52 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1NC2=NC(NC3=CC=C(N4CCC(N5CCN(CC5)C)CC4)C=C3OC)=NC=C2Cl)(C(C)C)=O
Molecular Weight (g/mol) 614.2
Formula Weight 614.2
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only