missing translation for 'onlineSavingsMsg'
Learn More

NVP-TAE 684, MedChemExpress

Product Code. p-200099548
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490027

Brand: Medchem Express HY10192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-TAE 684 (TAE 684) is a highly potent and selective ALK inhibitor, which blocks the growth of ALCL-derived and ALK-dependent cell lines with IC50 values between 2 and 10 nM.

NVP-TAE 684 (TAE 684) is a highly potent and selective ALK inhibitor, which blocks the growth of ALCL-derived and ALK-dependent cell lines with IC50 values between 2 and 10 nM.

Specifications

Chemical Name or Material NVP-TAE 684
CAS 761439-42-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H40ClN7O3S
Quantity 50 mg
Synonym TAE 684
Solubility Information DMSO : 7.69 mg/mL (12.52 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1NC2=NC(NC3=CC=C(N4CCC(N5CCN(CC5)C)CC4)C=C3OC)=NC=C2Cl)(C(C)C)=O
Molecular Weight (g/mol) 614.2
Formula Weight 614.2
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.