missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534868
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is one of a family of nuclear RNA export factor genes. It encodes a protein that is involved in mRNA export, is located in the nucleoplasm, and is associated with the nuclear envelope. Alternative splicing seems to be a common mechanism in this gene family. Two variants have been found for this gene.
Specifications
| NXF2 | |
| Polyclonal | |
| Unconjugated | |
| NXF2 | |
| 4930455N06Rik; cancer/testis antigen 39; CT39; Nuclear RNA export factor 2; nuclear RNA export factor 5; NXF2; NXF2B; Nxf-b; RGD1559530; TAPL2; TAPL-2; TAP-like protein 2; t-complex protein 11 homolog; TCP11X2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56001 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9GZY0 | |
| NXF2 | |
| Recombinant protein encompassing a sequence within the center region of human NXF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction