missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598024
This item is not returnable.
View return policy
Description
NXN Polyclonal Antibody for Western Blot, ELISA
This gene encodes a member of the thioredoxin superfamily, a group of small, multifunctional redox-active proteins. Members of this family are characterized by a conserved active motif called the thioredoxin fold that catalyzes disulfide bond formation and isomerization. The encoded protein acts a redox-dependent regulator of the Wnt signaling pathway and is involved in cell growth and differentiation.
Specifications
| NXN | |
| Polyclonal | |
| Unconjugated | |
| NXN | |
| Gn25; l11Jus13; NRX; nucleoredoxin; nucleoredoxin 1; NXN; Protein Red-1; TRG-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64359 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6DKJ4 | |
| NXN | |
| Recombinant Human Nucleoredoxin protein (155-435AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction