missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXNL2 Antibody (OTI2B11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73134CL1
This item is not returnable.
View return policy
Description
NXNL2 Monoclonal antibody specifically detects NXNL2 in Human,Monkey,Mouse,Primate samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| NXNL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| C9orf121, chromosome 9 open reading frame 121, nucleoredoxin-like 2, nucleoredoxin-like protein 2, RDCVF2 | |
| Full length human recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey, Mouse, Primate | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI2B11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 158046 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction