missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXNL2 Antibody (OTI2B11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73134DL594
This item is not returnable.
View return policy
Description
NXNL2 Monoclonal antibody specifically detects NXNL2 in Human, Mouse, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NXNL2 | |
| Monoclonal | |
| DyLight 594 | |
| C9orf121, chromosome 9 open reading frame 121, nucleoredoxin-like 2, nucleoredoxin-like protein 2, RDCVF2 | |
| Full length recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Monkey, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2B11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 158046 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction