missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NXPH1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15722071 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15722071 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15722071 Supplier Invitrogen™ Supplier No. PA554134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NLTNGGKSEL LKSGSSKSTL KHIWTESSKD LSISRLLSQT FRGKENDTDL DLRYDTPEPY SEQDLWDWLR NSTDLQEPRP RAKRRPIVKT GKFKKMFGW Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.

NXPH1 is a secreted protein with a variable N-terminal domain, a highly conserved, N-glycosylated central domain, a short linker region, and a cysteine-rich C-terminal domain. This protein forms a very tight complex with alpha neurexins, a group of proteins that promote adhesion between dendrites and axons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXPH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NXPH1
Gene Accession No. P58417
Gene Alias C130005L03Rik; Nbla00697; neurexophilin 1; neurexophilin-1; Neurophilin; Nph1; NXPH1
Gene Symbols NXPH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NXPH1. Recombinant protein control fragment (Product #RP-92694).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.