Learn More
Novus Biologicals™ NXT2 Overexpression Lysate
Code produit 18140240 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantity:
0.1 mg
Conditionnement:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
18140240 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18140240 Fournisseur Novus Biologicals™ Code fournisseur NBP206763
  • ONLINE EXCLUSIVE PRICE
    € 751.00
    € 709.80 / 0.1mg
    Save € 41.20
    5% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
The NXT2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_61168. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens NXT2, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native