missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NY-ESO-1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5116201

Product Code. 17137438

  • € 508.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NY-ESO-1 Polyclonal Antibody for Western Blot, IHC (P)

NY-ESO-1, a member of the CT (cancer/testis) family, is a tumor specific shared antigen with distinctive immunogenicity. The molecular basis of T cell-mediated antitumor immunity has been elucidated by the identification of a number of tumor antigens recognized by CD8+ T cells. Among these antigens, NY-ESO-1 is of particular interest because both the cytotoxic T lymphocyte and antibody have been shown to react with this antigen. The NY-ESO-1 has been detected in normal testis and in a range of human tumor types. NY-ESO-1 reactivity in testis is restricted germ cells, particularly in the spermatogonia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NY-ESO-1
Polyclonal
Unconjugated
CTAG1A
Autoimmunogenic cancer/testis antigen NY-ESO-1; cancer/testis antigen 1; cancer/testis antigen 1A; cancer/testis antigen 1-A; cancer/testis antigen 6.1; CT6.1; CTAG; CTAG1; CTAG1A; CTAG1B; ESO1; l antigen family member 2; LAGE2; LAGE-2; LAGE2A; LAGE2B; New York Esophageal Squamous Cell Carcinoma 1; NY-ESO-1
Rabbit
Antigen affinity chromatography, Protein A
RUO
246100
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.46 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P78358
CTAG1A
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CTAG1A.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.