Learn More
Invitrogen™ NYAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140085
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YDAVHSGSLS RSSPSVPHST PRPVSQDGAK MVNAAVNTYG AAPGGSRSRT PTSPLEELTS LFSSGRSLLR KSSSG Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 92%.
Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
| NYAP2 | |
| Polyclonal | |
| Unconjugated | |
| NYAP2 | |
| 9430031J16Rik; Jr6; Kiaa1486; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2; NEWGENE_1305560; NYAP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57624 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P242 | |
| NYAP2 | |
| Recombinant protein corresponding to Human NYAP2. Recombinant protein control fragment (Product #RP-108934). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.