Learn More
Masterflex™ Nylon Barbed Elbow Pipe Adapter
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30623-01
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12695710
  • € 25.85 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
Pipe Adapter Elbow
Not Autoclavable
1 21/32 in.
1/4 in.
1/4 in.
Translucent
Barb x NPT(M) Connection
Nylon
1/2 in.
Ethylene Oxide
1 21/32 in.