Learn More
Cole-Parmer™ Nylon Manifold, 2 outlets
Expand your flow and line capacity
Brand: Cole-Parmer™ 31522-33
Additional Details : Weight : 0.80000kg
Description
- Constructed of nylon for economic general applications.
- Offers good chemical compatibility.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
180°F | |
200 psi | |
2 3/8 in. | |
Nylon Manifold, 2 Outlets | |
-54°C | |
3/8 in. NPT(F) | |
3.18 cm |
82°C | |
Nylon | |
6.03 cm | |
-60°F | |
3/8 in. | |
1/4 in. NPT(F) | |
3.18 cm |