missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon NPT(F) union elbows
Chemically-inert PFA handles most aggressive chemicals
Brand: Masterflex™ 06349-60
Additional Details : Weight : 0.80000kg
Description
- Nylon NPT(F) union elbows
- Available in a variety of matierials and sizes
- Pack of Six
Specifications
Nylon | |
23 mm | |
6 Fittings | |
Ethylene Oxide | |
Elbow-shaped |
0.9 in. | |
PFA NPT(F) Union Elbows | |
1/8 in. NPT(F) | |
23 mm |