Learn More
Masterflex™ Nylon Straight Barbed Union
Specifications
Specifications
| Diameter (Metric) Inner | 12.7 mm |
| Material | Nylon |
| For Use With | 12.7mm |
| Min. Operating Temperature (Metric) | -46°C |
| Packaging Quantity | 10 Pack |
| Max. Operating Temperature (Metric) | 121°C |
| Product Type | Straight Barbed Union |