Learn More
Brady™ Nylon Thermal Transfer Printer Labels
For smooth, textured/rough or Highly textured/Low surface energy surfaces.
Brand: Brady™ 217067
Additional Details : Weight : 0.21750kg
Description
- Permanent acrylic adhesive
- Matt finish
- Nylon cloth labels
Specifications
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) | |
Acrylic, Permanent | |
Matte | |
Thermal Transfer | |
Laboratory identification | |
12.53 mm | |
3000/Pk. | |
Round | |
40 to 90°C | |
13.53 mm |
Thermal Transfer Printable Labels | |
Yellow | |
Nylon | |
Thermal Transfer Printer Label | |
BBP11, PR200Plus, PR300Plus, PR600Plus, BP1244, BP1344, BP4000, BP4320 | |
3 | |
3000/Pk. | |
9.53 mm dia. | |
0.041 mm | |
9.53 mm |