missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NYREN18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582752
This item is not returnable.
View return policy
Description
Immunogen sequence: DNRQESPSQE NIDRLVYMGF DALVAEAALR VFRGNVQLAA QTLAHNGGSL PPELPLSPED SLSPPATSPS DSAGTSSAST DEDMETEAVN EILEDIPEHE EDYLDSTLED EE Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 86%.
NYREN18 is a specific down-regulator of the NEDD8 conjugation system. It recruits NEDD8 and its conjugates to the proteasome for degradation. It also interacts with PSMD4/S5a, a member of the regulatory subunit of the 26S proteasome.
Specifications
| NYREN18 | |
| Polyclonal | |
| Unconjugated | |
| NUB1 | |
| 4931404D21Rik; 6330412F12Rik; BS4; EGK_14356; NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; negative regulator of ubiquitin like proteins 1; negative regulator of ubiquitin-like proteins 1; Nub1; NUB1L; NYREN18; NY-REN-18; NY-REN-18 antigen; Protein BS4; Renal carcinoma antigen NY-REN-18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5A7 | |
| NUB1 | |
| Recombinant protein corresponding to Human NUB1. Recombinant protein control fragment (Product #RP-92608). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction