missing translation for 'onlineSavingsMsg'
Learn More

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99+%

Product Code. 15415275
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415275 5 g 5g
15425275 25 g 25g
15435275 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15415275 Supplier Thermo Scientific Alfa Aesar Supplier No. H54732.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 330641-16-2 | C11H15BClF4N5O | 355.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330641-16-2
Molecular Formula C11H15BClF4N5O
Molecular Weight (g/mol) 355.53
MDL Number MFCD04973270
InChI Key GBGVQFJZGHBZMC-UHFFFAOYSA-N
Synonym tctu, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-6-chloro-1h-benzotriazol-1-yl uronium tetrafluoroborate, o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborat e, 6-chlorobenzotriazol-1-yl oxy-dimethylamino methylene-dimethyl-ammonium tetrafluoroborate, tctu; 1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium tetrafluoroborate, 6-chloro-1,2,3-benzotriazol-1-yl oxy dimethylamino methylidene dimethylazanium tetrafluoroborate, 1h-benzotriazolium, 1-bis dimethylamino methylene-5-chloro-, 3-oxide tetrafluoroborate 1-1:1, acmc-1agto, ksc569q6h
PubChem CID 24898642
IUPAC Name [(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C

Specifications

Melting Point 198°C to 204°C
Quantity 5 g
Formula Weight 355.53
Percent Purity ≥99%
Chemical Name or Material O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.