missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl Ganciclovir N-Formamide Diacetate, TRC

Product Code. 30337832
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337832 2.5 mg 2.5mg
30333271 5 mg 5mg
30328959 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30337832 Supplier Toronto Research Chemicals Supplier No. TRCB4123452.5MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Benzyl Ganciclovir N-Formamide Diacetate
Analyte or Component Names O-Benzyl Ganciclovir N-Formamide Diacetate
Shipping Condition Room Temperature
Molecular Formula C21 H23 N5 O7
InChI Formula InChI=1S/C21H23N5O7/c1-14(28)30-9-17(10-31-15(2)29)33-13-26-11-22-18-19(26)24-21(23-12-27)25-20(18)32-8-16-6-4-3-5-7-16/h3-7,11-12,17H,8-10,13H2,1-2H3,(H,23,24,25,27)
Solution Type Neat
SMILES CC(=O)OCC(COC(=O)C)OCn1cnc2c(OCc3ccccc3)nc(NC=O)nc12
IUPAC Name [3-acetyloxy-2-[(2-formamido-6-phenylmethoxypurin-9-yl)methoxy]propyl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 457.437
Quantity 2.5 mg
Formula Weight 457.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.