missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl Ganciclovir N-Formyl Diacetate Dimer, TRC

Product Code. 30341025
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341025 5 mg 5mg
30340533 2.5 mg 2.5mg
30331221 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30341025 Supplier Toronto Research Chemicals Supplier No. TRCB4123405MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Benzyl Ganciclovir N-Formyl Diacetate Dimer
Analyte or Component Names O-Benzyl Ganciclovir N-Formyl Diacetate Dimer
Shipping Condition Room Temperature
Molecular Formula C43 H46 N10 O14
InChI Formula InChI=1S/C43H46N10O14/c1-28(56)60-17-34(18-61-29(2)57)66-26-50-21-44-36-38(50)46-42(48-40(36)64-15-32-11-7-5-8-12-32)52(24-54)23-53(25-55)43-47-39-37(41(49-43)65-16-33-13-9-6-10-14-33)45-22-51(39)27-67-35(19-62-30(3)58)20-63-31(4)59/h5-14,21-22,24-25,34-35H,15-20,23,26-27H2,1-4H3
Solution Type Neat
SMILES CC(=O)OCC(COC(=O)C)OCn1cnc2c(OCc3ccccc3)nc(nc12)N(CN(C=O)c4nc(OCc5ccccc5)c6ncn(COC(COC(=O)C)COC(=O)C)c6n4)C=O
IUPAC Name [3-acetyloxy-2-[[2-[[[9-(1,3-diacetyloxypropan-2-yloxymethyl)-6-phenylmethoxypurin-2-yl]-formylamino]methyl-formylamino]-6-phenylmethoxypurin-9-yl]methoxy]propyl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 926.884
Quantity 5 mg
Formula Weight 926.319
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.