missing translation for 'onlineSavingsMsg'
Learn More

O-De(3-fluorobenzyl) Lapatinib Ditosylate Salt, TRC

Product Code. 30011599
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011599 2.5 mg 2.5mg
30133219 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011599 Supplier Toronto Research Chemicals Supplier No. TRCD2287502.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C36H37ClN4O10S3
Molecular Weight (g/mol) 817.35
Synonym 2-Chloro-4-[[6-[5-[[[2-(methylsulfonyl)ethyl]amino]methyl]-2-furanyl]-4-quinazolinyl]amino]phenol Ditosylate;
SMILES CC1=CC=C(S(=O)(O)=O)[C]=C1.OC(C(Cl)=C2)=CC=C2NC3=NC=NC4=CC=C(C5=CC=C(CNCCS(=O)(C)=O)O5)C=C43

Specifications

Analyte or Component Names O-De(3-fluorobenzyl) Lapatinib Ditosylate Salt
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 2.5 mg
Chemical Name or Material O-De(3-fluorobenzyl) Lapatinib Ditosylate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.