missing translation for 'onlineSavingsMsg'
Learn More

O-Desbromo-pyrimidinyl Macitentan, TRC

Product Code. 30349493
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349493 10 mg 10mg
30357109 25 mg 25mg
30366290 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30349493 Supplier Toronto Research Chemicals Supplier No. TRCD28202510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393813-43-8
Molecular Formula C15H19BrN4O4S
Molecular Weight (g/mol) 431.3
Synonym N-[5-(4-Bromophenyl)-6-[2-hydroxyethoxy]-4-pyrimidinyl]-N'-propylsulfamide
SMILES CCCNS(=O)(=O)Nc1ncnc(OCCO)c1c2ccc(Br)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H19BrN4O4S/c1-2-7-19-25(22,23)20-14-13(11-3-5-12(16)6-4-11)15(18-10-17-14)24-9-8-21/h3-6,10,19,21H,2,7-9H2,1H3,(H,17,18,20)
Formula Weight 430.031
Chemical Name or Material O-Desbromo-pyrimidinyl Macitentan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.