missing translation for 'onlineSavingsMsg'
Learn More

O-(Diphenylphosphinyl)hydroxylamine, 96%, Thermo Scientific Chemicals

Catalog Number p-7084043
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
72804-96-7
C12H12NO2P
233.21
SJECIYLGISUNRO-UHFFFAOYSA-N
o-diphenylphosphinylhydroxylamine, aminooxy diphenylphosphine oxide, o-diphenylphosphinyl hydroxylamine, o-diphenylphosphoryl hydroxylamine, amino diphenylphosphinate, hydroxylamine, o-diphenylphosphinyl, unii-2o9pf20t75, aminooxy diphenyl phosphine oxide, o-diphenylphosphinyl hydroxylamine mi
10955453
O-diphenylphosphorylhydroxylamine
NOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
72804-96-7
C12H12NO2P
233.21
SJECIYLGISUNRO-UHFFFAOYSA-N
o-diphenylphosphinylhydroxylamine, aminooxy diphenylphosphine oxide, o-diphenylphosphinyl hydroxylamine, o-diphenylphosphoryl hydroxylamine, amino diphenylphosphinate, hydroxylamine, o-diphenylphosphinyl, unii-2o9pf20t75, aminooxy diphenyl phosphine oxide, o-diphenylphosphinyl hydroxylamine mi
10955453
O-diphenylphosphorylhydroxylamine
NOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104.0°C
Assay Percent Range 88% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 233.21
Percent Purity 96%
Chemical Name or Material O-(Diphenylphosphinyl)hydroxylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'