missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAc Antibody (9B9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59309
This item is not returnable.
View return policy
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| O-GlcNAc | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| Mouse | |
| PEG purified | |
| RUO | |
| Specific for B-O-Linked N-Acetylgalactosamine and B-O-Linked N-Acetylglucosamine modified proteins. Does not cross-react with Glucose or Galactose modified proteins. | |
| Store at -20C. | |
| IgM |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 9B9 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| GlcNAc, O-linked N-acetylglucosamine | |
| Synthetic B-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction