missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAc Antibody (RL2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80892
This item is not returnable.
View return policy
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Flow (Intracellular), CyTOF-ready.
Specifications
| O-GlcNAc | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot : 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 20404350), Flow Cytometry : 1:10 - 1:1000, ELISA 1:100 - 1:2000,. Use reported in scientific literature (PMID 12029848), Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation : 1:10 - 1:500, Immunohistochemistry-Paraffin : 1:200, Dot Blot : 1:800, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready | |
| GlcNAc, O-linked N-acetylglucosamine | |
| Pore complex-lamina fraction purified from rat liver nuclear envelopes. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus species | |
| Purified |
| Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| RL2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Porcine reactivity reported in scientific literature (PMID: 26004176). Xenopus reactivity reported in scientific literature (PMID: 17329255). Please note that this antibody is reactive to Mouse and derived from the same host, Mouse. Additional Mouse on Mouse blocking steps may be required for IHC and ICC experiments. Please contact Technical Support for more information. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction