missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAc Antibody (RL2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-524H
This item is not returnable.
View return policy
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| O-GlcNAc | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus species | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RL2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| GlcNAc, O-linked N-acetylglucosamine | |
| Pore complex-lamina fraction purified from rat liver nuclear envelopes. | |
| 0.1 mL | |
| Detects nuclear pore complex (NPC), cytoplasmic and intranuclear O-linked glycoproteins from human, mouse, and rat tissues. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction