missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAcase/OGA/MGEA5 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32233G
This item is not returnable.
View return policy
Description
O-GlcNAcase/OGA/MGEA5 Polyclonal antibody specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| The immunogen this antibody was made to, maps to a region between residue 500 to 550 of human Meningioma Expressed Antigen 5 using the numbering given in entry NP_036347.1 (GeneID 10724). | |
| 0.1 mL | |
| 10724 | |
| Store at 4°C in the dark. |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction