Learn More
O-(tert-Butyl)hydroxylamine hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 39684-28-1 |
| Molecular Formula | C4H12ClNO |
| MDL Number | MFCD00043272 |
| Quantity | 25 g |
| SMILES | Cl.CC(C)(C)ON |
| Molecular Weight (g/mol) | 125.60 |
| Percent Purity | 98% |
| Chemical Name or Material | O-(tert-Butyl)hydroxylamine hydrochloride |