Learn More
O-tert-Butyl-L-serine methyl ester hydrochloride, 98%
Product Code. 15467066
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15467066
Brand: Thermo Scientific Alfa Aesar H62126.06
Request Bulk or Custom Format
CAS: 17114-97-5 | C8H18ClNO3 | 211.69 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17114-97-5
Molecular Formula C8H18ClNO3
Molecular Weight (g/mol) 211.69
MDL Number MFCD00077108
InChI Key PCIABNBULSRKSU-UHFFFAOYNA-N
Synonym h-ser tbu-ome.hcl, o-tert-butyl-l-serine methyl ester hydrochloride, h-ser tbu-ome hcl, s-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, methyl o-tert-butyl-l-serinate hydrochloride, l-serine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, methyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride, ksc491o3r, h-ser tbu-och3 hydrochloride, s-methyl 2-amino-3-tert-butoxy propanoate hcl
PubChem CID 16218549
SMILES Cl.COC(=O)C(N)COC(C)(C)C
Specifications
Beilstein 2041979
IUPAC Name methyl (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride
Quantity 5 g
Formula Weight 211.69
Percent Purity 98%
Chemical Name or Material O-tert-Butyl-L-serine methyl ester hydrochloride