Learn More
o-Thiocresol, 98%
Código de producto. 11470627
Cambiar vista
Click to view available options
Quantity:
5 g
25 g
100 g
Tamaño de la unidad:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
11470627 5 g 5g
11480627 25 g 25g
11498700 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11470627 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor B22460.06
Request Bulk or Custom Format
Este artículo no se puede devolver. Vea la política de devoluciones
CAS: 137-06-4 | C7H8S | 124.20 g/mol
o-Thiocresol is used as a thiol for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Thiocresol is used as a thiol for proteomics research.
Solubility
Soluble in ether and ethanol. Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 137-06-4
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004838
InChI Key LXUNZSDDXMPKLP-UHFFFAOYSA-N
Synonym o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
PubChem CID 8712
IUPAC Name 2-methylbenzenethiol
SMILES CC1=CC=CC=C1S
Especificaciones
Melting Point 10°C to 12°C
Density 1.054
Boiling Point 195°C
Flash Point 63°C (145°F)
Refractive Index 1.58
Quantity 5 g
UN Number UN2810
Beilstein 774068
Sensitivity Air Sensitive
Merck Index 14,9325
Solubility Information Soluble in ether and ethanol. Insoluble in water.
Formula Weight 124.21
Percent Purity 98%
Chemical Name or Material o-Thiocresol
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • o-Thiocresol
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: o-Thiocresol
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.